Thermo Scientific Chemicals

Praseodymium(III,IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194620500
Each
Thermo Scientific Chemicals

Praseodymium(III,IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 194620500
Each
Catalog number: 194620500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12037-29-5
IUPAC Name
hexapraseodymium(3+) undecaoxidandiide
Molecular Formula
O11Pr6
InChI Key
HPZIIFVSYNLWNX-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Appearance (Color)
Dark green to dark brown to black
Appearance (Form)
Powder
Total conc. of imp.as REO
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text