Thermo Scientific Chemicals

Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals

Catalog number: 194670250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals

Catalog number: 194670250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194670250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24424-99-5
IUPAC Name
di-tert-butyl dicarbonate
Molecular Formula
C10H18O5
InChI Key
DYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
Appearance (Color)
White
Appearance (Form)
Low melting crystalline solid
Infrared spectrum
Conforms
Titration by coupling
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text