Di-tert-butyl dicarbonate, 99%
Di-tert-butyl dicarbonate, 99%
Di-tert-butyl dicarbonate, 99%
Di-tert-butyl dicarbonate, 99%
Thermo Scientific Chemicals

Di-tert-butyl dicarbonate, 99%

CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
50 g
10 g
100 g
Packaging:
Glass bottle
Catalog number 194675000
Price (USD)
440.65
Special offer
Online exclusive
Ends: 26-Sep-2025
544.00
Save 103.35 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
440.65
Special offer
Online exclusive
Ends: 26-Sep-2025
544.00
Save 103.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS24424-99-5
IUPAC Namedi-tert-butyl dicarbonate
Molecular FormulaC10H18O5
InChI KeyDYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)OC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Titration by coupling98.5 to 101.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469151Certificate of AnalysisJul 09, 2025194670500, 194671000, 194670250, 194670100, 194675000
A0462433Certificate of AnalysisJun 20, 2025194670500, 194671000, 194670250, 194670100, 194675000
A0473012Certificate of AnalysisJun 16, 2025194670500, 194671000, 194670250, 194670100, 194675000
A0469151Certificate of AnalysisJan 31, 2025194670500, 194671000, 194670250, 194670100, 194675000
A0458201Certificate of AnalysisJan 31, 2025194670500, 194671000, 194670250, 194670100, 194675000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link