Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 194700025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 194700025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 194700025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
546-68-9
IUPAC Name
titanium(4+) tetrakis(propan-2-olate)
Molecular Formula
C12H28O4Ti
InChI Key
VXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES
[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Appearance (Form)
Clear liquid
Color scale
=<100 APHA
Appearance
May darken on storage
Infrared spectrum
Conforms
Melting point
>=15°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text