Lithium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Lithium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 194715000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol)160.43
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Insoluble matter=<0.005 % (in water)
Chloride (Cl)=<50 ppm
Appearance (Form)Crystalline or granular powder or crystals
Titration after Ion exchange99.0 to 101.0 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
RUO – Research Use Only