Thermo Scientific Chemicals

Lithium perchlorate trihydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 194720010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium perchlorate trihydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 194720010
1 kg, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 194720010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-78-6
IUPAC Name
lithium(1+) trihydrate perchlorate
Molecular Formula
ClH6LiO7
InChI Key
JAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES
[Li+].O.O.O.[O-][Cl](=O)(=O)=O
Titration Argentometric
>=98.5 %
Chloride (Cl)
=<30 ppm
Sulfate (SO4)
=<10 ppm
Iron (Fe)
=<5 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text