Tellurium(IV) chloride, 99%
Tellurium(IV) chloride, 99%
Tellurium(IV) chloride, 99%
Tellurium(IV) chloride, 99%
Thermo Scientific Chemicals

Tellurium(IV) chloride, 99%

CAS: 10026-07-0 | Cl4Te | 269.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 194740250
Price (USD)
107.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
107.00
Each
Add to cart
Chemical Identifiers
CAS10026-07-0
IUPAC Nametetrachloro-λ⁴-tellane
Molecular FormulaCl4Te
InChI KeySWLJJEFSPJCUBD-UHFFFAOYSA-N
SMILESCl[Te](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Powder
Titration Argentometric>=98.5 % (ex Cl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection