Thermo Scientific Chemicals

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Catalog number: 194780500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dimethyl aminoterephthalate, 99%, Thermo Scientific Chemicals

Catalog number: 194780500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 194780500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5372-81-6
IUPAC Name
1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate
Molecular Formula
C10H11NO4
InChI Key
DSSKDXUDARIMTR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C(=O)OC)C(N)=C1
Appearance (Form)
Powder or crystals
Appearance (Color)
Yellow to beige to light green to brown
Melting point
131°C to 134°C
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text