Thermo Scientific Chemicals

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Catalog number: 194795000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Catalog number: 194795000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 194795000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-46-0
IUPAC Name
trimethyl[(trimethylsilyl)oxy]silane
Molecular Formula
C6H18OSi2
InChI Key
UQEAIHBTYFGYIE-UHFFFAOYSA-N
SMILES
C[Si](C)(C)O[Si](C)(C)C
Appearance (Form)
Liquid
Refractive index
1.3765 to 1.3785 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text