Thermo Scientific Chemicals

May-Gruenwald Stain, pure

Catalog number: 194830250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

May-Gruenwald Stain, pure

Catalog number: 194830250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194830250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
524698-43-9
Appearance (Color)
Dark green to dark brown
UV Lambda max
655 to 670 nm (blue)
Ratio
1.2 to 2.1 Blue/red ratio
Appearance (Form)
Crystalline powder
UV Lambda max
in 66:33 IMS:water

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text