Thermo Scientific Chemicals

Dibutyltin dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 194872500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Dibutyltin dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 194872500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194872500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
683-18-1
Molecular Formula
C8H20Cl2Sn
InChI Key
GSCBVGBIRKBMAC-UHFFFAOYSA-N
SMILES
Cl.Cl.CCCC[SnH2]CCCC
Molecular Weight (g/mol)
305.86
Appearance (Color)
White to beige
Appearance (Form)
Crystals or fused crystalline mass
Infrared spectrum
Conforms
Titration Argentometric
95 to 105 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text