Thermo Scientific Chemicals

Copper(I) iodide, 98%, Thermo Scientific Chemicals

Catalog number: 194900010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Copper(I) iodide, 98%, Thermo Scientific Chemicals

Catalog number: 194900010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 194900010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7681-65-4
IUPAC Name
λ¹-copper(1+) iodide
Molecular Formula
CuI
InChI Key
LSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES
[Cu+].[I-]
Sulfate (SO4)
=<5000 ppm
Titration Complexometric
>=97.5 %
Appearance (Form)
Powder
Appearance (Color)
Off-white to grey-brown
Iron (Fe)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text