Copper(I) iodide, 98%
Copper(I) iodide, 98%
Copper(I) iodide, 98%
Copper(I) iodide, 98%
Thermo Scientific Chemicals

Copper(I) iodide, 98%

CAS: 7681-65-4 | CuI | 190.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 194902500
Price (USD)
100.65
Special offer
Online exclusive
Ends: 26-Sep-2025
124.00
Save 23.35 (19%)
Each
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
100.65
Special offer
Online exclusive
Ends: 26-Sep-2025
124.00
Save 23.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7681-65-4
IUPAC Nameλ¹-copper(1+) iodide
Molecular FormulaCuI
InChI KeyLSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES[Cu+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<5000 ppm
Titration Complexometric>=97.5 %
Appearance (Form)Powder
Appearance (Color)Off-white to grey-brown
Iron (Fe)=<50 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection