Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Ammonium metavanadate, 99.5%, for analysis
Thermo Scientific Chemicals

Ammonium metavanadate, 99.5%, for analysis

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 194910500
Price (USD)
34.65
Special offer
Online exclusive
Ends: 26-Sep-2025
42.80
Save 8.15 (19%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 26-Sep-2025
42.80
Save 8.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS7803-55-6
IUPAC Namevanadiumoylol amine
Molecular FormulaH4NO3V
InChI KeyYBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILESN.O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Copper (Cu)=<10 ppm
Appearance (Form)Crystalline powder
Cobalt (Co)=<20 ppm
Lead (Pb)=<20 ppm
Iron (Fe)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link