Thermo Scientific Chemicals

Tetraethylammonium acetate tetrahydrate, 97+%, Thermo Scientific Chemicals

Catalog number: 194940250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetraethylammonium acetate tetrahydrate, 97+%, Thermo Scientific Chemicals

Catalog number: 194940250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194940250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
67533-12-4
IUPAC Name
tetraethylazanium acetate tetrahydrate
Molecular Formula
C10H31NO6
InChI Key
QNOAKQGNSWGYOE-UHFFFAOYSA-M
SMILES
O.O.O.O.CC([O-])=O.CC[N+](CC)(CC)CC
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
41°C to 46°C
Titration with HClO4
97.0 to 103.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text