Sodium borodeuteride, 98 atom% D, Thermo Scientific Chemicals
Sodium borodeuteride, 98 atom% D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium borodeuteride, 98 atom% D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 194950010
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
Add to cart
Sodium borodeuteride, 98 atom% D, Thermo Scientific Chemicals
Catalog number194950010
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.10 
Save 10.45 (14%)
-
Add to cart
Chemical Identifiers
CAS15681-89-7
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
D-Enrichment>=97.5 %
Appearance (Form)Powder and/or chunks
Appearance (Color)White to off-white to light grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only