Thermo Scientific Chemicals

1-Methylcyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 194960050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Methylcyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 194960050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 194960050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6914-76-7
IUPAC Name
1-methylcyclopropane-1-carboxylic acid
Molecular Formula
C5H8O2
InChI Key
DIZKLZKLNKQFGB-UHFFFAOYSA-N
SMILES
CC1(CC1)C(O)=O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text