Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Diethylene glycol dibutyl ether, 99+%, pure
Thermo Scientific Chemicals

Diethylene glycol dibutyl ether, 99+%, pure

CAS: 112-73-2 | C12H26O3 | 218.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 194980010
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS112-73-2
IUPAC Name1-[2-(2-butoxyethoxy)ethoxy]butane
Molecular FormulaC12H26O3
InChI KeyKZVBBTZJMSWGTK-UHFFFAOYSA-N
SMILESCCCCOCCOCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity (CH3COOH)=<0.01 %
GC>=99.0 %
Refractive index1.4225 to 1.4245 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<15 APHA
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link