Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific Chemicals
Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 194980025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-73-2
IUPAC Name1-[2-(2-butoxyethoxy)ethoxy]butane
Molecular FormulaC12H26O3
InChI KeyKZVBBTZJMSWGTK-UHFFFAOYSA-N
SMILESCCCCOCCOCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Acidity (CH3COOH)=<0.01 %
GC>=99.0 %
Refractive index1.4225 to 1.4245 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<15 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only