Search Thermo Fisher Scientific
Description | MSA-330 Multi-Stream Slurry XRF Analyzer |
Certifications/Compliance | IP66 electrical rating. Manufactured in an ISO 9001:2000 certified facility. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
19502 | Each | MSA-330 Multi-Stream Slurry XRF Analyzer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The MSA-330 quantifies % solids and up to 20 elements for between 3 and 12 slurry streams according to a predefined schedule to deliver real-time process trend information for monitoring and control. Key features include: