Thermo Scientific Chemicals

Potassium tetrachloropalladate(II), min 32.0% Pd, Thermo Scientific Chemicals

Catalog number: 195120020
2 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium tetrachloropalladate(II), min 32.0% Pd, Thermo Scientific Chemicals

Catalog number: 195120020
2 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195120020
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-98-6
IUPAC Name
dipotassium tetrachloropalladiumdiuide
Molecular Formula
Cl4H4K2Pd
InChI Key
OANHYYMESYMMDY-UHFFFAOYSA-N
SMILES
Cl.Cl.Cl.Cl.[K+].[K+].[Pd--]
Magnesium (Mg)
=<10 ppm
Ruthenium (Ru)
=<20 ppm
Rhodium (Rh)
=<20 ppm
Iridium (Ir)
=<20 ppm
Platinum (Pt)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text