Palladium black, 98% Pd, surface area 28-33 m2/g, Thermo Scientific Chemicals
Palladium black, 98% Pd, surface area 28-33 m2/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium black, 98% Pd, surface area 28-33 m2/g, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 195150050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
Molecular Weight (g/mol)106.42
View more
SpecificationsSpecification SheetSpecification Sheet
Palladium (Pd)>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only