Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals
Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Packaging:
Glass bottle
Catalog number 195170020
Price (USD)/ Each
-
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepalladium(2+) diammonium tetrachloride
Molecular FormulaCl4H8N2Pd
InChI KeyIRKVTUVFHGUMMN-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]
Molecular Weight (g/mol)284.30
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Palladium (Pd)>=36.5 %
Appearance (Color)Green to red to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only