Thermo Scientific Chemicals

Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals

Catalog number: 195170100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium tetrachloropalladate(II), 98%, Thermo Scientific Chemicals

Catalog number: 195170100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195170100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13820-40-1
IUPAC Name
palladium(2+) diammonium tetrachloride
Molecular Formula
Cl4H8N2Pd
InChI Key
IRKVTUVFHGUMMN-UHFFFAOYSA-L
SMILES
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]
Appearance (Form)
Powder or crystals
Palladium (Pd)
>=36.5 %
Appearance (Color)
Green to red to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text