Thermo Scientific Chemicals

Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals

Catalog number: 195180100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals

Catalog number: 195180100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195180100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3375-31-3
IUPAC Name
palladium(2+) diacetate
Molecular Formula
C4H6O4Pd
InChI Key
YJVFFLUZDVXJQI-UHFFFAOYSA-L
SMILES
[Pd++].CC([O-])=O.CC([O-])=O
Infrared spectrum
Conforms
Iron (Fe)
=<50 ppm
Chromium (Cr)
=<10 ppm
Lead (Pb)
=<50 ppm
Magnesium (Mg)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text