Invitrogen™

Biotin-14-dCTP

Catalog number: 19518018
Have Questions?
Invitrogen™

Biotin-14-dCTP

Catalog number: 19518018
Have Questions?
Catalog Number
19518018
Unit Size
50 nmol
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1951801850 nmol
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Biotin-14-dCTP is used for DNA biotinylation, an excellent, and sometimes superior alternative to conventional radioisotopic labeling. Advantages of using biotinylated DNA probes include:

• Convenience and safety—probe preparation generates no radioactive waste
• Stability—probes remain active longer than isotopically labeled probes
• Versatility—biotinylated DNA can be used for immobilization and capture procedures via streptavidin binding

About Biotin-14-dCTP
Biotin-14-dCTP is a patented dCTP analog with biotin attached at the N4-position of the pyrimidine base by a 14-atom linker. This nucleotide can be efficiently incorporated into DNA by nick translation in the presence of dATP, dGTP, and dTTP, labeling DNA with terminal deoxynucleotide transferase, or random primer synthesis. The amount of material provided is sufficient for labeling up to 50 μg of DNA.

Performance and quality testing
Purity is evaluated by reverse-phase HPLC.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Labeled Nucleotide
Concentration
0.4 mM
Label or Dye
Biotin & Other Hapten
Labeling Method
Direct Labeling
Shipping Condition
Dry Ice
Quantity
50 nmol
Form
Liquid

Contents & Storage

Biotin-14-dCTP is supplied at 0.4 mM in 100 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text