Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals
Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
2 g
10 g
Packaging:
View all
Glass Bottle
Catalog number 195180500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS3375-31-3
IUPAC Namepalladium(2+) diacetate
Molecular FormulaC4H6O4Pd
InChI KeyYJVFFLUZDVXJQI-UHFFFAOYSA-L
SMILES[Pd++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Iron (Fe)=<50 ppm
Chromium (Cr)=<10 ppm
Lead (Pb)=<50 ppm
Magnesium (Mg)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only