Platinum black, 98% Pt, Thermo Scientific Chemicals
Platinum black, 98% Pt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum black, 98% Pt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 195210050
Price (USD)/ Each
862.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
862.00
Add to cart
Platinum black, 98% Pt, Thermo Scientific Chemicals
Catalog number195210050
Price (USD)/ Each
862.00
-
Add to cart
Chemical Identifiers
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
Molecular Weight (g/mol)195.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Specific surface area>=25 m²/g
Appearance (Form)Powder
Assay>=97.5 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only