Platinum on activated carbon, 5% Pt, Thermo Scientific Chemicals
Platinum on activated carbon, 5% Pt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum on activated carbon, 5% Pt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 195230100
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
Add to cart
Platinum on activated carbon, 5% Pt, Thermo Scientific Chemicals
Catalog number195230100
Price (USD)/ Each
128.65
Online exclusive
143.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Additional infototal surface area (m²/g): 900
Additional info< 7 um: 10%
Additional infotypical properties:
Additional info< 28 um: 50%
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only