Invitrogen™

Biotin-14-dATP

Catalog number: 19524016
Have Questions?
Invitrogen™

Biotin-14-dATP

Catalog number: 19524016
Have Questions?
Catalog Number
19524016
Unit Size
50 nmol
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1952401650 nmol
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Biotin-14-dATP is a patented dATP analog with biotin attached at the 6-position of the purine base by a 14-atom linker. The long, 14-atom spacer arm allows accessibility to streptavidin for sensitive detection of probe-target hybrids in in situ and filter-based hybridizations. Biotin-14-dATP can be efficiently incorporated into DNA by nick translation in the presence of dCTP, dGTP, and dTTP (1). Biotin-14-dATP can be added to the 3´ end of DNA using terminal deoxynucleotidyl transferase (2). The amount supplied is sufficient for labeling up to 50 μg of DNA by nick translation.

Performance and Quality Testing: Purity is evaluated by reverse-phase HPLC analysis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Liquid
Labeling Method
Direct Labeling
Label or Dye
Biotin & Other Hapten
Product Type
Labeled Nucleotide
Quantity
50 nmol
Shipping Condition
Approved for shipment on Wet or Dry Ice
Concentration
0.4 mM

Contents & Storage

Biotin-14-dATP is supplied at 0.4 mM in 100 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text