Thermo Scientific Chemicals

Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals

Catalog number: 195240500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals

Catalog number: 195240500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195240500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-06-4
IUPAC Name
platinum
Molecular Formula
Pt
InChI Key
BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES
[Pt]
Appearance (Color)
Black
Platinum (Pt)
9.8 to 10.2 %
Appearance (Form)
Powder
Moisture
45 % to 60 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text