Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals
Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 mg
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number 195380250
Price (USD)/ Each
2,103.65
Online exclusive
2475.00 
Save 371.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
2,103.65
Online exclusive
2475.00 
Save 371.35 (15%)
Add to cart
Platinum(II) chloride, 73% Pt, Thermo Scientific Chemicals
Catalog number195380250
Price (USD)/ Each
2,103.65
Online exclusive
2475.00 
Save 371.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
Molecular Weight (g/mol)265.98
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey-green to brown
Assay>=71.8 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only