1x1 image pixel for data collection
Platinum(II) chloride, 73% Pt
Platinum(II) chloride, 73% Pt
Platinum(II) chloride, 73% Pt
Platinum(II) chloride, 73% Pt
Thermo Scientific Chemicals

Platinum(II) chloride, 73% Pt

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Packaging:
Glass bottle
Catalog number 195382500
Price (USD)
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
71.30
Save 13.65 (19%)
Each
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
71.30
Save 13.65 (19%)
Each
Add to cart
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey-green to brown
Assay>=71.8 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link