Thermo Scientific Chemicals

Platinum(IV) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 195400010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Platinum(IV) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 195400010
1 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 195400010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13454-96-1
IUPAC Name
tetrachloroplatinum
Molecular Formula
Cl4Pt
InChI Key
FBEIPJNQGITEBL-UHFFFAOYSA-J
SMILES
Cl[Pt](Cl)(Cl)Cl
Lead (Pb)
=<25 ppm
Silicon (Si)
=<200 ppm
Trace metal analysis
=<500 ppm (Sum Ru+Rh+Ir+Pd)
Appearance (Color)
Brown
Iron (Fe)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text