Platinum(IV) chloride, 99%
Platinum(IV) chloride, 99%
Platinum(IV) chloride, 99%
Platinum(IV) chloride, 99%
Thermo Scientific Chemicals

Platinum(IV) chloride, 99%

CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 195402500
Price (USD)
61.00
Each
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.00
Each
Add to cart
Chemical Identifiers
CAS13454-96-1
IUPAC Nametetrachloroplatinum
Molecular FormulaCl4Pt
InChI KeyFBEIPJNQGITEBL-UHFFFAOYSA-J
SMILESCl[Pt](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<25 ppm
Silicon (Si)=<200 ppm
Trace metal analysis=<500 ppm (Sum Ru+Rh+Ir+Pd)
Appearance (Color)Brown
Iron (Fe)=<50 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link