Thermo Scientific Chemicals

Rhodium on activated carbon, 5% Rh, unreduced, dry, Thermo Scientific Chemicals

Catalog number: 195440050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Rhodium on activated carbon, 5% Rh, unreduced, dry, Thermo Scientific Chemicals

Catalog number: 195440050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195440050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-16-6
IUPAC Name
rhodium
Molecular Formula
Rh
InChI Key
MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES
[Rh]
Appearance (Color)
Grey-black to black
Assay
4.5 to 5.5 % (Rh)
Particle Size/Sieve analysis
< 8 um: 21%
Particle Size/Sieve analysis
<128 um: 99%
Particle Size/Sieve analysis
< 16 um: 39%

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text