Thermo Scientific Chemicals

Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals

Catalog number: 195480010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Ruthenium(III) chloride hydrate, 35 - 40% Ru, Thermo Scientific Chemicals

Catalog number: 195480010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195480010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14898-67-0
IUPAC Name
ruthenium(3+) trichloride
Molecular Formula
Cl3Ru
InChI Key
YBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ru+3]
Sodium (Na)
=<300 ppm
Gold (Au)
=<300 ppm
Iron (Fe)
=<300 ppm
Potassium (K)
=<300 ppm
Osmium (Os)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text