Celestine Blue, pure, Thermo Scientific Chemicals
Celestine Blue, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Celestine Blue, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 195560500
Price (USD)/ Each
290.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
342.00 
Save 51.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
290.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
342.00 
Save 51.35 (15%)
Add to cart
Celestine Blue, pure, Thermo Scientific Chemicals
Catalog number195560500
Price (USD)/ Each
290.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
342.00 
Save 51.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name{amino[7-(diethylamino)-3,4-dioxo-4,10-dihydro-3H-phenoxazin-1-yl]methylidene}oxidanium chloride
Molecular FormulaC17H18ClN3O4
InChI KeyREPMZEQSQQAHJR-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC=C2NC3=C(OC2=C1)C(=O)C(=O)C=C3C(N)=[OH+]
Molecular Weight (g/mol)363.80
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to dark brown to black
Appearance (Form)Powder
Infrared spectrumConforms
UV spectrumComplies
UV Lambda max638 to 648 nm (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only