Thermo Scientific Chemicals

D(+)-Raffinose pentahydrate, 99+%

Catalog number: 195671000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

D(+)-Raffinose pentahydrate, 99+%

Catalog number: 195671000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195671000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17629-30-0
IUPAC Name
(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C18H32O16
InChI Key
MUPFEKGTMRGPLJ-ZQSKZDJDSA-N
SMILES
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Appearance (Color)
White
Sulfated ash
=<0.1 % (on dry basis)
Specific optical rotation
+103.0° to +107.0° (20°C, 589 nm) (c=10, water)
Solubility
(10 % in water) Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text