Thermo Scientific Chemicals

Carbol Fuchsin, pure

Catalog number: 195790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Carbol Fuchsin, pure

Catalog number: 195790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 195790250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4197-24-4
IUPAC Name
hydrogen 4-[(4-aminophenyl)[(1Z)-4-imino-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]-2-methylaniline chloride
Molecular Formula
C21H22ClN3
InChI Key
HZLHRDBTVSZCBS-GHTYLULLSA-N
SMILES
[H+].[Cl-].CC1=CC(=CC=C1N)C(\C1=CC=C(N)C=C1)=C1\C=CC(=N)C(C)=C1
UV spectrum
Complies
UV Lambda max
(in water) about 542 nm
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Suitability
(for microscopy) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text