Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
Catalog number195881000
Price (USD)/ Each
26.20
-
Add to cart
Specifications
Chemical Name or MaterialCalcium standard solution
Concentration997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C)
Name Note1 mg/ml Ca in 0.5N HNO3
CAS7440-70-2
PackagingPlastic bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.