Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, Thermo Scientific Chemicals
Catalog number196001000
Price (USD)/ Each
57.20
-
Add to cart
Specifications
Chemical Name or MaterialGold standard solution
Concentration990 to 1010μg/mL of Au(3+) in 10 to 20% HCl
Name Note1 mg/ml Au in 0.5N HCl
CAS7440-57-5
PackagingPlastic bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.