Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Catalog Number
1960533
Unit Size
Each
Description
CO2 Backup System
Type
CO2 Backup System
For Use With (Equipment)
Revco™ ExF, DxF, CxF Series
Price (USD)
Availability
-
Full specifications
Quantity
-
Description
CO2 Backup System
Type
CO2 Backup System
Temperature Range
-80°C
For Use With (Equipment)
Revco™ ExF, DxF, CxF Series, TSE and TSD ULT Series
LN2 backup system maintains temperature down to -80°C with liquid nitrogen
CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.