Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Thermo Scientific Chemicals

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 196055000
Price (USD)
82.65
Special offer
Online exclusive
Ends: 26-Sep-2025
101.00
Save 18.35 (18%)
Each
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
82.65
Special offer
Online exclusive
Ends: 26-Sep-2025
101.00
Save 18.35 (18%)
Each
Add to cart
Specifications
CAS7439-89-6
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:82664
Chemical Name or MaterialIron standard solution
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link