Thermo Scientific Chemicals

Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals

Catalog number: 196111000
100 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Manganese standard solution, for AAS, 1mg/mL Mn in 2 to 5% HNO3, Thermo Scientific Chemicals

Catalog number: 196111000
100 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196111000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7439-96-5, 7697-37-2, 7732-18-5
Appearance (Form)
Solution
Strength
990 to 1010 µg/ml of Mn(2+) in 2 to 5% HNO3
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text