Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals
Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
Plastic bottle
Catalog number 196171000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium standard solution
Concentration997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C)
Name Note1 mg/ml Pd in 10% HCl
CAS7440-05-3
PackagingPlastic bottle
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only