Thermo Scientific Chemicals

Ruthenium standard solution, for AAS, 1mg/mL Ru in 5% HCl, Thermo Scientific Chemicals

Catalog number: 196251000
100 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Ruthenium standard solution, for AAS, 1mg/mL Ru in 5% HCl, Thermo Scientific Chemicals

Catalog number: 196251000
100 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196251000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-18-8, 7647-01-0, 7732-18-5
Appearance (Form)
Solution
Appearance (Color)
Clear red-brown
Strength
990 to 1010 ug/ml of Ru(3+) in 10-20% HCl

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text