Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals
Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Plastic bottle
Catalog number 196285000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7782-49-2, 7647-01-0, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Strength995 to 1005 ug/ml of Se in 10% HCl (20°C)
Appearance (Form)Clear solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only