Thermo Scientific Chemicals

Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, Thermo Scientific Chemicals

Catalog number: 196301000
100 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, Thermo Scientific Chemicals

Catalog number: 196301000
100 mL, Plastic bottle, Each
Quantity
Packaging
Catalog number: 196301000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7761-88-8, 7697-37-2, 7732-18-5
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Strength
997 to 1003 ug/ml of Ag (1+) in 0.5N HNO3 (20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text