1x1 image pixel for data collection
MabCaptureC Affinity Matrix
MabCaptureC Affinity Matrix
Thermo Scientific™

MabCaptureC Affinity Matrix

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1963662250250 mL
196366201L1 L
196366205L5 L
196366210L10 L
Catalog number 1963662250
Price (USD)
-
Quantity:
250 mL

MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

MabCaptureC™ Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.

Features of the MabCaptureC™ Affinity Matrix include:

  • High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Operating pressure: ≤3 bar
  • Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
  • Formulation buffer: 20% (v/v) ethanol

For process development and resin screening, MabCaptureC™ is also available in the following formats:

  • MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
  • MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
  • MabCaptureC™ RoboColumn, 200 μL (5943662200)
  • MabCaptureC™ RoboColumn, 600 μL (5943662600)
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Binding Capacity>50 g IgG/L
For Use With (Application)Monoclonal antibody purification
FormAgarose Bead
FormulationSuspension in 20% ethanol
LigandEngineered rProtein A
MatrixAgarose-based, epoxide-activated
Particle Size75 ±10 μm
Quantity250 mL
TargetCH2-CH3 interface Fc-region
Product TypeAffinity Matrix
Unit SizeEach

Share catalog number, name or link